accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3OONNHH3CCH3CH3OONNHOOOHHO

CAS RN 1041026-71-4

IUPAC Name: oxalic acid; bis(tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate)
Synonyms: oxalic acid; bis(tert-butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate)
Molecular Weight (g/mol): 486.57
Molecular Formula: C22H38N4O8
InChi Key: PYRAMVPOMDHTNX-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.CC(C)(C)OC(=O)N1CC2(CNC2)C1.CC(C)(C)OC(=O)N1CC2(CNC2)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)