accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOOOCH3

CAS RN 104104-12-3

IUPAC Name: 1-methoxy-4-{2-[2-(4-methoxyphenoxy)ethoxy]ethoxy}benzene
Synonyms: 1-methoxy-4-{2-[2-(4-methoxyphenoxy)ethoxy]ethoxy}benzene
Molecular Weight (g/mol): 318.37
Molecular Formula: C18H22O5
InChi Key: AJXHXSKQHBJNPB-UHFFFAOYSA-N
SMILES: COC1=CC=C(OCCOCCOC2=CC=C(OC)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results