accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CClClOHO

CAS RN 10411-52-6

IUPAC Name: 2,3-dichloro-2-methylpropanoic acid
Synonyms: sodium 2,3-dichloroisobutyrate
Molecular Weight (g/mol): 156.99
Molecular Formula: C4H6Cl2O2
InChi Key: LRGIIZXVSULULZ-UHFFFAOYNA-N
SMILES: CC(Cl)(CCl)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results