accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CON

CAS RN 10412-98-3

IUPAC Name: 5-oxohexanenitrile
Synonyms: 5-oxohexanenitrile
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: AEVMBQIIZGKQRB-UHFFFAOYSA-N
SMILES: CC(=O)CCCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results