accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104147-69-5

IUPAC Name: 1-(3-bromopropoxy)-2-nitrobenzene
Synonyms: 1-(3-bromopropoxy)-2-nitrobenzene
Molecular Weight (g/mol): 260.09
Molecular Formula: C9H10BrNO3
InChi Key: HPZBIRIHQKSGGT-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1OCCCBr
Boiling Point 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results