accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104163-35-1

IUPAC Name: 1-(3-methylthiophen-2-yl)methanamine
Synonyms: 1-(3-methylthiophen-2-yl)methanamine
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: SWZNXCABBUKIPZ-UHFFFAOYSA-N
SMILES: CC1=C(CN)SC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results