accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10419-77-9

IUPAC Name: diethyl (iodomethyl)phosphonate
Synonyms: diethyl iodomethylphosphonate
Molecular Weight (g/mol): 278.03
Molecular Formula: C5H12IO3P
InChi Key: PNFHTBDRPOJJTQ-UHFFFAOYSA-N
SMILES: CCOP(=O)(CI)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results