CAS RN 104202-15-5
IUPAC Name:
1-bromo-3-phenylimidazo[1,5-a]pyridine
Synonyms:
1-bromo-3-phenylimidazo[1,5-a]pyridine
Molecular Weight (g/mol):
273.13
Molecular Formula:
C13H9BrN2
InChi Key:
ULKZZBGVTIZXSA-UHFFFAOYSA-N
SMILES:
BrC1=C2C=CC=CN2C(=N1)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Bromo-3-phenylimidazo[1,5-a]pyridine 97.0+%, TCI America™
CAS: 104202-15-5 Molecular Formula: C13H9BrN2 Molecular Weight (g/mol): 273.13 MDL Number: MFCD04587511 InChI Key: ULKZZBGVTIZXSA-UHFFFAOYSA-N PubChem CID: 957540 IUPAC Name: 1-bromo-3-phenylimidazo[1,5-a]pyridine SMILES: BrC1=C2C=CC=CN2C(=N1)C1=CC=CC=C1