accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClFNOO

CAS RN 104222-34-6

IUPAC Name: 5-chloro-4-fluoro-2-nitroaniline
Synonyms: 5-chloro-4-fluoro-2-nitroaniline
Molecular Weight (g/mol): 190.56
Molecular Formula: C6H4ClFN2O2
InChi Key: VRJKEIWZSOHDOH-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=C(F)C=C1[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)