accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHHOO

CAS RN 10425-11-3

IUPAC Name: 4-benzoylbenzene-1,2-diol
Synonyms: 4-benzoylbenzene-1,2-diol
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: ARWCZKJISXFBGI-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1O)C(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results