accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OONNOCH3

CAS RN 104296-35-7

IUPAC Name: ethyl 5-(furan-2-yl)-1-methyl-1H-pyrazole-3-carboxylate
Synonyms: ethyl 5-(furan-2-yl)-1-methylpyrazole-3-carboxylate
Molecular Weight (g/mol): 220.23
Molecular Formula: C11H12N2O3
InChi Key: QEOVLTVAGMDVLQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=NN(C)C(=C1)C1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results