accessibility menu, dialog, popup

UserName

Viewing profile as user:
KKOSeOO

CAS RN 10431-47-7

IUPAC Name: dipotassium selenite
Synonyms: dipotassium selenite
Molecular Weight (g/mol): 205.17
Molecular Formula: K2O3Se
InChi Key: RNGFNLJMTFPHBS-UHFFFAOYSA-L
SMILES: [K+].[K+].[O-][Se]([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results