accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNO

CAS RN 10431-98-8

IUPAC Name: 2-ethyl-4,5-dihydro-1,3-oxazole
Synonyms: polyethyloxazoline
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: NYEZZYQZRQDLEH-UHFFFAOYSA-N
SMILES: CCC1=NCCO1
Boiling Point 
  • (3)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)