accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10431-99-9

IUPAC Name: 2-(propan-2-yl)-4,5-dihydro-1,3-oxazole
Synonyms: 2-isopropyl-4,5-dihydro-1,3-oxazole
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: FVEZUCIZWRDMSJ-UHFFFAOYSA-N
SMILES: CC(C)C1=NCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results