accessibility menu, dialog, popup

UserName

Viewing profile as user:
OONH2HCl

CAS RN 104347-13-9

IUPAC Name: 3-aminooxolan-2-one hydrochloride
Synonyms: HSLs hydrochloride
Molecular Weight (g/mol): 137.56
Molecular Formula: C4H8ClNO2
InChi Key: XBKCXPRYTLOQKS-UHFFFAOYNA-N
SMILES: Cl.NC1CCOC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results