accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSONCH

CAS RN 10442-03-2

IUPAC Name: 4-(prop-2-yn-1-yl)-1λ⁶-thiomorpholine-1,1-dione
Synonyms: 4-(prop-2-yn-1-yl)-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol): 173.23
Molecular Formula: C7H11NO2S
InChi Key: CZBZIZOYSYHBNM-UHFFFAOYSA-N
SMILES: O=S1(=O)CCN(CC#C)CC1
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)