accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHN

CAS RN 10443-61-5

IUPAC Name: 2-undecyl-4,5-dihydro-1H-imidazole
Synonyms: 2-undecyl-4,5-dihydro-1H-imidazole
Molecular Weight (g/mol): 224.39
Molecular Formula: C14H28N2
InChi Key: FQHUDZKKDCTQET-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=NCCN1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results