accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10443-70-6

IUPAC Name: ethyl 4-(4-chloro-2-methylphenoxy)butanoate
Synonyms: mcpb-ethyl
Molecular Weight (g/mol): 256.73
Molecular Formula: C13H17ClO3
InChi Key: XNKARWLGLZGMGX-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCOC1=CC=C(Cl)C=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results