accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SNNFFF

CAS RN 10444-89-0

IUPAC Name: 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
Synonyms: 5-(trifluoromethyl)-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol): 169.13
Molecular Formula: C3H2F3N3S
InChi Key: LTEUXHSAYOSFGQ-UHFFFAOYSA-N
SMILES: NC1=NN=C(S1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)