CAS RN 104458-24-4
IUPAC Name:
2-methanesulfonylethan-1-amine hydrochloride
Synonyms:
2-methanesulfonylethanamine hydrochloride
Molecular Weight (g/mol):
159.63
Molecular Formula:
C3H10ClNO2S
InChi Key:
AMYYUKGKCJKCBI-UHFFFAOYSA-N
SMILES:
Cl.CS(=O)(=O)CCN
1
–
1
of
1
results
1
2-Aminoethyl Methyl Sulfone Hydrochloride 98.0+%, TCI America™
CAS: 104458-24-4 Molecular Formula: C3H10ClNO2S Molecular Weight (g/mol): 159.628 MDL Number: MFCD03840162 InChI Key: AMYYUKGKCJKCBI-UHFFFAOYSA-N Synonym: 2-(Methylsulfonyl)ethanamine Hydrochloride PubChem CID: 17749886 IUPAC Name: 2-methylsulfonylethanamine;hydrochloride SMILES: CS(=O)(=O)CCN.Cl