accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104508-27-2

IUPAC Name: 3-(phenoxymethyl)benzaldehyde
Synonyms: 3-(phenoxymethyl)benzaldehyde
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: KZUJGQAFTCDBKA-UHFFFAOYSA-N
SMILES: O=CC1=CC(COC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results