CAS RN 1046-56-6
5,6-Diphenyl-3-(2-pyridyl)-1,2,4-triazine 98.0+%, TCI America™
CAS: 1046-56-6 Molecular Formula: C20H14N4 Molecular Weight (g/mol): 310.36 MDL Number: MFCD00006462 InChI Key: OTMYLOBWDNFTLO-UHFFFAOYSA-N Synonym: 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine,5,6-diphenyl-3-2-pyridyl-1,2,4-triazine,1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl,5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine,as-triazine, 5,6-diphenyl-3-2-pyridyl,5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine,maybridge1_007490,bio7c7,acmc-1boy4,cbmicro_025706 PubChem CID: 70588 IUPAC Name: 5,6-diphenyl-3-(pyridin-2-yl)-1,2,4-triazine SMILES: C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1
5,6-Diphenyl-3-(2-pyridyl)-1,2 4-triazine, 99%
CAS: 1046-56-6 Molecular Formula: C20H14N4 Molecular Weight (g/mol): 310.36 MDL Number: MFCD00006462 InChI Key: OTMYLOBWDNFTLO-UHFFFAOYSA-N Synonym: 3-2-pyridyl-5,6-diphenyl-1,2,4-triazine,5,6-diphenyl-3-2-pyridyl-1,2,4-triazine,1,2,4-triazine, 5,6-diphenyl-3-2-pyridinyl,5,6-diphenyl-3-pyridin-2-yl-1,2,4-triazine,as-triazine, 5,6-diphenyl-3-2-pyridyl,5,6-diphenyl-3-2-pyridinyl-1,2,4-triazine,maybridge1_007490,bio7c7,acmc-1boy4,cbmicro_025706 PubChem CID: 70588 SMILES: C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1
Sigma Aldrich 3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.