CAS RN 1046153-20-1
IUPAC Name:
2-oxa-6-azaspiro[3.5]nonane
Synonyms:
2-oxa-6-azaspiro[3.5]nonane
Molecular Weight (g/mol):
127.19
Molecular Formula:
C7H13NO
InChi Key:
SDESQEIYRGXXPA-UHFFFAOYSA-N
SMILES:
C1OCC11CCCNC1
1
–
1
of
1
results
1
Sigma Aldrich 2-Oxa-6-azaspiro[3.5]nonane oxalate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.