accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10466-65-6

IUPAC Name: potassium rheniumoylolate
Synonyms: potassium tetraoxorhenate(1-)
Molecular Weight (g/mol): 289.3
Molecular Formula: KO4Re
InChi Key: QFKRWIFGDGKWLM-UHFFFAOYSA-N
SMILES: [K+].[O-][Re](=O)(=O)=O
Molecular Weight (g/mol) 
  • (4)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (6)
  • (1)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (4)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)
  • (3)
  • (1)