accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10466-72-5

IUPAC Name: 6-[(2,4-dinitrophenyl)amino]hexanoic acid
Synonyms: dnp-cap dnp-epsilon-aminocaproate dnp-aminocaproic acid
Molecular Weight (g/mol): 297.27
Molecular Formula: C12H15N3O6
InChi Key: ZYUWUKIAUDIXCQ-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results