accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104663-56-1

IUPAC Name: manganese(3+) phosphate
Synonyms: manganese(3+) phosphate
Molecular Weight (g/mol): 149.91
Molecular Formula: MnO4P
InChi Key: BECVLEVEVXAFSH-UHFFFAOYSA-K
SMILES: [Mn+3].[O-]P([O-])([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results