accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104678-18-4

IUPAC Name: (2-methoxyethyl)(propan-2-yl)amine
Synonyms: isopropyl(2-methoxyethyl)amine
Molecular Weight (g/mol): 117.19
Molecular Formula: C6H15NO
InChi Key: UMCVTLHNNUEZDO-UHFFFAOYSA-N
SMILES: COCCNC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results