accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10469-09-7

IUPAC Name: tetrachloropyridine-2-carboxylic acid
Synonyms: tetrachloropyridine-2-carboxylic acid
Molecular Weight (g/mol): 260.88
Molecular Formula: C6HCl4NO2
InChi Key: GXFRQLQUKBSYQL-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(Cl)=C(Cl)C(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results