accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 104706-47-0

IUPAC Name: pyrrolidin-3-ol hydrochloride
Synonyms: pyrrolidin-3-ol hydrochloride
Molecular Weight (g/mol): 123.58
Molecular Formula: C4H10ClNO
InChi Key: QPMSJEFZULFYTB-UHFFFAOYNA-N
SMILES: Cl.OC1CCNC1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results