accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOOCH3

CAS RN 10471-28-0

IUPAC Name: 1,12-diethyl dodecanedioate
Synonyms: 1,12-diethyl dodecanedioate
Molecular Weight (g/mol): 286.41
Molecular Formula: C16H30O4
InChi Key: AFYNPRWLOKYLDU-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCCCCCCCCC(=O)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results