accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H2COO

CAS RN 10475-46-4

IUPAC Name: naphthalen-2-yl 2-methylprop-2-enoate
Synonyms: naphthalen-2-yl 2-methylprop-2-enoate
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: CXOYJPWMGYDJNW-UHFFFAOYSA-N
SMILES: CC(=C)C(=O)OC1=CC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results