accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10477-47-1

IUPAC Name: prop-2-yn-1-yl prop-2-enoate
Synonyms: prop-2-yn-1-yl prop-2-enoate
Molecular Weight (g/mol): 110.11
Molecular Formula: C6H6O2
InChi Key: WPBNLDNIZUGLJL-UHFFFAOYSA-N
SMILES: C=CC(=O)OCC#C
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)