CAS RN 10478-99-6
IUPAC Name:
2-(1,4-dihydropyridin-4-ylidene)-2,3-dihydro-1H-indene-1,3-dione
Synonyms:
2-(1H-pyridin-4-ylidene)indene-1,3-dione
Molecular Weight (g/mol):
223.23
Molecular Formula:
C14H9NO2
InChi Key:
RFJVOUZINWUTIG-UHFFFAOYSA-N
SMILES:
O=C1C2=CC=CC=C2C(=O)C1=C1C=CNC=C1
1
–
2
of
2
results
1
2-[4(1H)-Pyridinylidene]indan-1,3-dione, TCI America™
CAS: 10478-99-6 Molecular Formula: C14H9NO2 Molecular Weight (g/mol): 223.23 MDL Number: MFCD00142802,MFCD00444488 InChI Key: RFJVOUZINWUTIG-UHFFFAOYSA-N Synonym: 2-[4(1H)-Pyridylidene]indan-1,3-dione, gamma-Pyrophthalone PubChem CID: 5389994 IUPAC Name: 2-(1,4-dihydropyridin-4-ylidene)-2,3-dihydro-1H-indene-1,3-dione SMILES: O=C1C2=CC=CC=C2C(=O)C1=C1C=CNC=C1
2
Sigma Aldrich 2-(4(1H)-Pyridinylidene)-1H-indene-1,3(2H)-dione
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.