accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1048-08-4

Si

CAS RN 1048-08-4

IUPAC Name: tetraphenylsilane
Synonyms: silane, tetraphenyl-
Molecular Weight (g/mol): 336.51
Molecular Formula: C24H20Si
InChi Key: JLAVCPKULITDHO-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)