accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NOONH

CAS RN 1048039-15-1

IUPAC Name: N-(adamantan-1-yl)-4-oxo-1-pentyl-6-phenyl-1,4-dihydroquinoline-3-carboxamide
Synonyms: N-(adamantan-1-yl)-4-oxo-1-pentyl-6-phenylquinoline-3-carboxamide
Molecular Weight (g/mol): 468.64
Molecular Formula: C31H36N2O2
InChi Key: MSJISJDTJJYBFT-UHFFFAOYSA-N
SMILES: CCCCCN1C=C(C(=O)NC23CC4CC(CC(C4)C2)C3)C(=O)C2=CC(=CC=C12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results