accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NI

CAS RN 104830-06-0

IUPAC Name: 3-iodopyridin-2-amine
Synonyms: 3-iodopyridin-2-amine
Molecular Weight (g/mol): 220.01
Molecular Formula: C5H5IN2
InChi Key: UUDNBWSHTUFGDQ-UHFFFAOYSA-N
SMILES: NC1=NC=CC=C1I
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)