accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CO

CAS RN 10486-19-8

IUPAC Name: tridecanal
Synonyms: tridecanal
Molecular Weight (g/mol): 198.35
Molecular Formula: C13H26O
InChi Key: BGEHHAVMRVXCGR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC=O
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)