CAS RN 10488-87-6
IUPAC Name:
ethyl 2-methyl-3-oxo-3-phenylpropanoate
Synonyms:
ethyl 2-methyl-3-oxo-3-phenylpropanoate
Molecular Weight (g/mol):
206.24
Molecular Formula:
C12H14O3
InChi Key:
RAWGHPXIJSCHFZ-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(C)C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Ethyl 2-Benzoylpropionate 98.0+%, TCI America™
CAS: 10488-87-6 Molecular Formula: C12H14O3 Molecular Weight (g/mol): 206.24 MDL Number: MFCD00026882 InChI Key: RAWGHPXIJSCHFZ-UHFFFAOYNA-N Synonym: 2-Benzoylpropionic Acid Ethyl Ester PubChem CID: 139166 IUPAC Name: ethyl 2-methyl-3-oxo-3-phenylpropanoate SMILES: CCOC(=O)C(C)C(=O)C1=CC=CC=C1