accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CNOOCH3H3CCH3

CAS RN 1048970-17-7

IUPAC Name: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate
Molecular Weight (g/mol): 311.23
Molecular Formula: C16H30BNO4
InChi Key: IBLQMWKHENBVJE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results