CAS RN 104934-54-5
IUPAC Name:
3-octadecylthiophene
Synonyms:
3-octadecylthiophene
Molecular Weight (g/mol):
336.62
Molecular Formula:
C22H40S
InChi Key:
ARFJPHXJBIEWSZ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCC1=CSC=C1
1
–
1
of
1
results
1
3-Octadecylthiophene 98.0+%, TCI America™
CAS: 104934-54-5 Molecular Formula: C22H40S Molecular Weight (g/mol): 336.622 MDL Number: MFCD00191567 InChI Key: ARFJPHXJBIEWSZ-UHFFFAOYSA-N Synonym: 3-Stearylthiophene PubChem CID: 566857 IUPAC Name: 3-octadecylthiophene SMILES: CCCCCCCCCCCCCCCCCCC1=CSC=C1