accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3C(R)OCH3NH2OCH3CH3CH3

CAS RN 104944-18-5

IUPAC Name: tert-butyl (2R)-2-amino-3-methylbutanoate hydrochloride
Synonyms: tert-butyl (2R)-2-amino-3-methylbutanoate hydrochloride|H-D-VAL-OTBU HCL
Molecular Weight (g/mol): 209.71
Molecular Formula: C9H20ClNO2
InChi Key: AUIVQIHTTVPKFS-OGFXRTJISA-N
SMILES: Cl.CC(C)[C@@H](N)C(=O)OC(C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)