accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONN

CAS RN 104949-82-8

IUPAC Name: 4-(hexyloxy)benzene-1,2-dicarbonitrile
Synonyms: 4-(hexyloxy)benzene-1,2-dicarbonitrile
Molecular Weight (g/mol): 228.3
Molecular Formula: C14H16N2O
InChi Key: BMXSATBWGFIAPA-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC(C#N)=C(C=C1)C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results