accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 10496-15-8

IUPAC Name: 1-(hexyldisulfanyl)hexane
Synonyms: 1-(hexyldisulfanyl)hexane
Molecular Weight (g/mol): 234.46
Molecular Formula: C12H26S2
InChi Key: GJPDBURPGLWRPW-UHFFFAOYSA-N
SMILES: CCCCCCSSCCCCCC
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results