accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1049677-30-6

IUPAC Name: diethyl (12-sulfanyldodecyl)phosphonate
Synonyms: diethyl 12-sulfanyldodecylphosphonate
Molecular Weight (g/mol): 338.49
Molecular Formula: C16H35O3PS
InChi Key: JAVFXEXYSKLPHD-UHFFFAOYSA-N
SMILES: CCOP(=O)(CCCCCCCCCCCCS)OCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results