CAS RN 1049738-54-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ ERK Inhibitor
The ERK Inhibitor, also referenced under CAS 1049738-54-6, controls the biological activity of ERK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.