accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONNOH(E)(E)

CAS RN 105-11-3

IUPAC Name: N-[(1E,4E)-4-(hydroxyimino)cyclohexa-2,5-dien-1-ylidene]hydroxylamine
Synonyms: N-[(1E,4E)-4-(hydroxyimino)cyclohexa-2,5-dien-1-ylidene]hydroxylamine
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: LNHURPJLTHSVMU-CGXWXWIYSA-N
SMILES: ON=C1C=CC(C=C1)=NO
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)