accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COON

CAS RN 105-34-0

IUPAC Name: methyl 2-cyanoacetate
Synonyms: methyl cyanoacetate malonic methyl ester nitrile
Molecular Weight (g/mol): 99.09
Molecular Formula: C4H5NO2
InChi Key: ANGDWNBGPBMQHW-UHFFFAOYSA-N
SMILES: COC(=O)CC#N
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)