accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCl

CAS RN 105-39-5

IUPAC Name: ethyl 2-chloroacetate
Synonyms: ethyl chloroacetate
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: VEUUMBGHMNQHGO-UHFFFAOYSA-N
SMILES: CCOC(=O)CCl
Boiling Point 
  • (3)
  • (4)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (5)
Percent Purity 
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (2)
  • (7)
  • (3)
  • (4)
  • (2)
  • (2)
  • (6)
  • (5)
  • (2)
  • (9)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)