CAS RN 105-43-1
IUPAC Name:
3-methylpentanoic acid
Synonyms:
3-methyl valeric acid
Molecular Weight (g/mol):
116.16
Molecular Formula:
C6H12O2
InChi Key:
IGIDLTISMCAULB-UHFFFAOYNA-N
SMILES:
CCC(C)CC(O)=O