accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 105-44-2

IUPAC Name: (Z)-N-(4-methylpentan-2-ylidene)hydroxylamine
Synonyms: (Z)-N-(4-methylpentan-2-ylidene)hydroxylamine
Molecular Weight (g/mol): 115.18
Molecular Formula: C6H13NO
InChi Key: ZKALVNREMFLWAN-SREVYHEPSA-N
SMILES: CC(C)C\C(C)=N/O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results